Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 10μL Microvolume Filter Tips

Catalog No. 14222745 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-745 0.5 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-745 Supplier Axygen™ Supplier No. T300STKS

missing translation for 'validMainframeMsg'

Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, bevelled, nonsterile, sterile, non-filtered, and other features for specialized applications.

  • Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention and high yield
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 10 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Model T-300-STK-S
Certifications/Compliance USP 3127
Clean Claims DNase, Rnase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Micro Point
Quantity 480/Pk., 4800/Cs.
Product Type Pipette Tips
For Use With (Equipment) Gilson Pipetman P2, Gilson Pipetman P10
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.