Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 20 μL Ultra Micro Tips

Catalog No. 14222761 Shop All Axygen Products
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack

Catalog No. 14-222-761

Supplier: Axygen™ T400RS

Only null left
Add to Cart
Add to Cart

Maxymum Recovery™ allows recovery of costly samples with improved precision and accuracy

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • RNase-/DNase-free
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention

Order Info

  • Bulk: 1000 tips/pack, 20 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Sterility Sterile
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Micro Point
For Use With (Equipment) Eppendorf, Gilson Pipetman P2, Gilson Pipetman P10
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.