Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference
GREENER_CHOICE

Catalog No. 14222811 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Format:
Bulk
Rack
Refill
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Color Format
14-222-811 Non-sterile Clear Bulk
14-222-816 Non-sterile Clear Rack
14-222-817 Sterile Clear Rack
14-222-820 Non-sterile Yellow Bulk
14-222-815 Non-sterile Clear Refill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-222-811 Supplier Axygen™ Supplier No. TR222C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed to fit a wide variety of single and multi-channel pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

(This is the suggested replacement for T-205-WB-C-L)

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore

Order Info

Available in bulk, racked or stack rack formats.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model TR-222-C
Certifications/Compliance EN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, Quality Control Tested
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Bulk
Tip Style Beveled, Universal
Packaging 1,000 Tips/Pk., 20 Packs/Cs.
Quantity 1000 Tips/Pk., 20 Pk./Cs.
Product Type Pipette Tips
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Maxymum Recovery
Material Polypropylene
Graduations 10, 50, 100μL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.