Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 300 μL Universal Pipetter Tips

Catalog No. 14222751 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
14-222-751 Sterile Rack
14-222-746 Non-sterile Bulk
14-222-750 Non-sterile Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-222-751 Supplier Axygen™ Supplier No. T350CRS

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fine-point tips fits popular brands of pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Fine-Point

Order Info

  • Bulk: 1000 tips/pack, 240 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 300 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Model T-350-C-R-S
Certifications/Compliance EN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, Sterility compliant with ANSI/AAMI/ISO 11137 (SAL 10−6), Quality Control Tested
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Rack
Tip Style Micro Point, Universal
Quantity 96/Rack, 10 Racks/Pk., 5 Pk./Cs.
Product Type Pipette Tips
Includes Tips, Packs, Case, Rack
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.