Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 48-Well PCR Microplates
GREENER_CHOICE

Catalog No. 14222319 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-222-319 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-319 Supplier Axygen™ Supplier No. PCR48C
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specially designed to fit most thermal cyclers

  • Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, DNA-free
Lid Without Lid
Array 6 x 8
Color Clear
Material Polypropylene
No. of Wells 48
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.