Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 5 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 384 Tip Format
GREENER_CHOICE

Catalog No. 14222892 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
5 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-892 5 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-222-892 Supplier Axygen™ Supplier No. VT3845ULR
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Agilent/Velocity11 automation systems

Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

  • Clear polypropylene
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Available in nonsterile or sterile

Order Info

Packaging:10 Racks/Pack, 5 Packs/Case

TRUSTED_SUSTAINABILITY

Specifications

Description Non-sterile
Volume (Metric) 5 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.