Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 96-well PCR Microplates

Catalog No. 14222327 Shop All Axygen Products
Change view
Click to view available options
Skirt Style:
Full
Half
Sterility:
Non-sterile
Sterile
Barcode:
Includes Barcode
No Barcode
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Skirt Style Sterility Barcode
14-222-327 Full Sterile No Barcode
14-224-002 Half Non-sterile Includes Barcode
14-224-003 Full Non-sterile Includes Barcode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-222-327 Supplier Axygen™ Supplier No. PCR96FSCS
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.

Axygen™ 96 well elevated skirt bar coded PCR microplate is designed to fit ABI™ Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film. 

  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 96 Well Semi-Skirted PCR Plate
  • Single Notch Coner
  • Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance RNase-, DNase-free
Format Standard
Array 8 x 12
Color Clear
For Use With (Equipment) Standard 96 well blocks
Skirt Style Full
Material Polypropylene
No. of Wells 96
Sterility Sterile
Well Shape Round
For Use With (Application) PCR thermal transfer, RT-PCR applications
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.