Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Axygen™ Automation Tip
GREENER_CHOICE

Catalog No. 14222850 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
45 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-850 45 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-222-850 Supplier Corning™ Supplier No. FXF38445FPRS
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Corning™ Axygen™ Automation Tip features unsurpassed straightness to ensure that the tip enters the well smoothly, every time.

  • Tip format: 384 tip
  • Tip material: Clear polypropylene
  • RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Compatible head: AP-384
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
  • Aerosol-barrier filtered robotic pipet tips
  • Fine point improves accuracy and precision at lower sample volumes
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 45 μL
Volume (Metric) Dispensing 25 to 50 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
Packaging Racked
Quantity 10 Racks/Pk, 5 Pk/Cs
For Use With (Equipment) Beckman Coulter, Biomek FX, Biomek NX, Multimek
Product Line Axygen
Material Polyethylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.