Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ AxyMats™ 96 Round Well Compression Mat for PCR Microplates

Catalog No. 14222063 Shop All Axygen Products
Change view
Click to view available options
Material:
Silicone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
14-222-063 Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 14-222-063 Supplier Axygen™ Supplier No. CM96RD
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Suited to help reduce evaporation and ensure proper seal when used in conjunction with Axygen PCR Microplates and PlateMax™ sealing films

Axygen™ Compression AxyMatsTM are ideally suited to help reduce evaporation and ensure a proper seal when used in conjunction with Axygen PCR Microplates and PlateMaxTM sealing films. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.

  • For use with 96/384 well and PCR microplates
  • TRUSTED_SUSTAINABILITY

    Specifications

    Color Natural
    Material Silicone
    Sterility Sterile
    Autoclavable Autoclavable
    No. of Wells 96
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Product Type Compression Mat
    Well Shape Round
    Plate Compatibility 96/384-well PCR Microplate
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.