Learn More
Axygen™ AxyMats™ Pierceable Sealing Mats for 24 Well Deep Well Plates
Catalog No. 14224005 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14-224-005 Sterile
14-222-008 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-224-005 Supplier Axygen™ Supplier No. AM24SQS
Minimize evaporation in wide range of temperatures in PCR and storage applications.
  • Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications
  • AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved
  • They can be reused following a bleach wash and ethanol rinse protocol.
  • Mfr. No. AM-24-SQ-S
TRUSTED_SUSTAINABILITY
Specifications
Material Silicone
Sterility Sterile
Autoclavable Autoclavable
No. of Wells 24
No. per Case 5
No. per Pack 10
Product Type Sealing Mat
Plate Compatibility 24-well Deep Well Plate with Square Wells
WARNING: Cancer - www.P65Warnings.ca.gov