Learn More
Axygen™ AxyMats™ Sealing Mat for 48 Well Deep Well Plates
Catalog No. 14222019 Shop All Axygen Products
Change view
Click to view available options
Material:
Silicone
Catalog No. Material
14-222-019 Silicone
1 options
This item is not returnable. View return policy
Catalog No. 14-222-019 Supplier Axygen™ Supplier No. AM48
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 48
No. per Case 50/Cs.
No. per Pack 10/Pk.
Product Type Sealing Mat
Well Shape Rectangular
Plate Compatibility 48-well Rectangular Deep Well Microplate, 5 mL
WARNING: Cancer - www.P65Warnings.ca.gov