Learn More
Axygen™ AxyMats™ Sealing Mat for 96 Well PCR Microplates
Catalog No. 14222024 Shop All Axygen Products
Change view
Click to view available options
Material:
Silicone
Catalog No. Material
14-222-024 Silicone
1 options
This item is not returnable. View return policy
Catalog No. 14-222-024 Supplier Axygen™ Supplier No. AM96PCRRD
Add to Cart
Add to Cart
This item is not returnable. View return policy
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50/Cs.
No. per Pack 10/Pk.
Product Type Sealing Mat
Well Shape Round
Plate Compatibility 96-well PCR Microplate
WARNING: Cancer - www.P65Warnings.ca.gov