Learn More
Axygen™ Axypet™ Pro Pipettor Starter Kit
GREENER_CHOICE
Catalog No. 07202447 Shop All Corning Products
Change view
Click to view available options
Number of Channels:
1
Catalog No. Number of Channels
07-202-447 1
1 options
Catalog No. 07-202-447 Supplier Axygen™ Supplier No. APSTRKITP
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Corning™ Axygen™ Axypet™ Pro Pipettors are engineered to provide the highest levels of comfort, accuracy, and precision.
Axygen™ Axypet™ Pro Starter Kit contains 4 variable volume pipettors in the most popular and universal volumes (0.5-10 μL, 2-20 μL, 20-200 μL, and 100-1000 μL), pipet stand for 4 pipettors, and 3 racks with tips.
  • Cat. No. AP-10-P Axygen Axypet Pro Single-channel Pipettor, 0.5 to 10 μL
  • Cat. No. AP-200-P Axygen Axypet Pro Single-channel Pipettor, 20 to 200 μL
  • Cat. No. AP-1000-P Axygen Axypet Pro Single-channel Pipettor, 100 to 1000 μL
  • Cat. No. AP-USD4-P Universal linear stand for four single-channel pipettors
  • Cat. No. MR-10XT-R (10 μL) Axygen Multirack pipet tips
  • Cat. No. MR-200-R (200 μL) Axygen Multirack pipet tips
  • Cat. No. MR-1000XT-R (1000 μL) Axygen
'
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO8655
Autoclavable Autoclavable
Color Red/Yellow/Blue
Grip Type Ergonomic
Number of Channels 1
Product Line Axypet
Type Starter Kit
Finger Style Ergonomic
Includes 4 single-channel pipettors, Linear stand, Sample tips, Instruction manual, QC Certificate, In-lab calibration tool
Quantity 1 Kit
Warranty 3
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov