Axygen™ AxyPrep Mag™ FragmentSelect Kits Available on GSA/VA Contract for Federal Government customers only.

Next generation sequencing sample preparation simplified

Manufacturer: Axygen Scientific MAGFRAGI1

 View more versions of this product

Catalog No. 14-223-232

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Genomic DNA purification
    Format 96- and 384-well plate
    Includes AxyPrep™ Mag™ FragmentSelect-I paramagnetic bead solution
    No. of Reactions 15 to 30 Preps
    Product Type Kit
    Quantity 1mL,1mL
    Sufficient For 15 to 30 Preps
    Product Name Kit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit