Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ AxyPrep™ Mag PCR Normalizer Kit

Catalog No. 14223228 Shop All Axygen Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-223-228 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-223-228 Supplier Axygen™ Supplier No. MAGPCRNM1
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The AxyPrep MAG PCR Normalizer Kit utilizes a paramagnetic beads-based purification system for post-PCR clean-up. The beads have limited binding surface; by limiting the amount of beads added in a purification reaction, a pre-defined amount of DNA can be isolated based on the need.

PCR normalization is accomplished during the purification process, so that additional DNA quantification and dilution are not necessary. Time, labor and reagent cost are greatly saved with the AxyPrep Mag unique normalization purification chemistry. The protocol mainly consists of binding, washing and elution steps, and can be performed directly in the thermal cycling plate. The process can be automated with walk away solution for high throughput applications.

  • Dual action: Normalization and post PCR clean-up enables significant reduction in processing and faster turnaround times
  • Scalable: Tube, 96- and 384-well microplate formats
  • Manual and automation-compatible protocol
  • Compatible with several downstream applications
  • No centrifugation or filtration required
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
Product Type Nucleic Acid Purification Kit
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.