Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Clear Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
GREENER_CHOICE

Catalog No. 14223745 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
300 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-223-745 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-223-745 Supplier Axygen™ Supplier No. HTF300CHTRS5
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations

Axygen offers an alternative choice for your Hamilton™ STARline system.

These tips are:

  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Available in non-sterile, sterile, non-filtered, and filtered
  • Available as Clear or Conductive black polypropylene tips
  • Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 300 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Hanging Tip Rack
Tip Style Automation
Packaging 5 Racks/Pk, 12 Pks/Cs.
For Use With (Equipment) Hamilton MICROLAB Nimbus
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.