Learn More
Andwin Scientific AXYGEN MICROFUGE TUBE 2.0ML
SDP
Catalog No. NC0086054
Encompass_Preferred
The 2.0 mL MaxyClear boil-proof microtube offers secure storage for samples as the tubes remain closed during boiling and centrifugation. This product also has frosted sides and cap that can be written on, which permit users to easily organize and identify the tubes. With a 0.28mm piercing port in the center of the cap, the tubes can easily be pierced by a syringe needle. The tubes are free of RNase, DNase, and pyrogen.
Catalog No. NC0086054 Supplier Andwin Scientific Supplier No. MCT200CS
The 2.0 mL MaxyClear boil-proof microtube offers secure storage for samples as the tubes remain closed during boiling and centrifugation. This product also has frosted sides and cap that can be written on, which permit users to easily organize and identify the tubes. With a 0.28mm piercing port in the center of the cap, the tubes can easily be pierced by a syringe needle. The tubes are free of RNase, DNase, and pyrogen.