Learn More
Axygen™ Microtube Racks: 80-Well Axyracks
Catalog No. 14223355 Shop All Axygen Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
14-223-355 Black
1 options
Catalog No. 14-223-355 Supplier Axygen™ Supplier No. R80BK
Ideal for storage and handling of PCR/Micro/Screw cap tubes
  • Holds eighty 1.5/2.0mL micro and screw cap tubes
  • Polypropylene; resistant to most lab chemicals and solvents
  • For cryogenic storage of tubes
  • Conical bottom wells for superior stability of conical tubes
  • Each well numbered for ease of sample identification
  • Frosted side writing panel
  • Available in a variety of fluorescent and non-fluorescent colors
  • Packaging: 25 Racks/Cs.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Holds Tubes (1.5 to 2 mL)
Material Polypropylene
Positions 80
Quantity 25/Cs.
Product Type Microtube Rack