Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Multi Well Low Profile Reagent Reservoirs
GREENER_CHOICE

Catalog No. 14222404 Shop All Axygen Products
Change view
Click to view available options
Capacity (Metric):
32 mL
84 mL
Sterility:
Non-sterile
Sterile
Channels:
8
12
Packaging:
25/Cs.
Individually Wrapped; 25/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Channels Packaging
14-222-404 84 mL Non-sterile 12 25/Cs.
14-222-405 84 mL Sterile 12 Individually Wrapped; 25/Cs.
14-222-409 32 mL Non-sterile 8 25/Cs.
14-222-410 32 mL Sterile 8 Individually Wrapped; 25/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-222-404 Supplier Axygen™ Supplier No. RESMW12LP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 84 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 25/Cs.
No. per Pack 25/Pk.
Packaging 25/Cs.
Volume (Metric) Well 7 mL
Channels 12
Disposable Single-use
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.