Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Multimek Robotic Tips: 200μL

Catalog No. 14222936 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14-222-936 Non-sterile
14-222-937 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 14-222-936 Supplier Axygen™ Supplier No. SCT200SSBRS
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Fits Multimek Type II or CCS-style head

Robotic tip's unique diamond-polished molding technology uses no mold-releasing agents. Minimizes wastage of precious samples and reagents while handling. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.

  • Tip volume: 200μL
  • Tip color: Clear
  • Rack color: Red and blue

Order Info

Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Blue
Clean Claims DNase, RNase, and Pyrogen free
For Use With (Application) Unique diamond polished molding technology use no mold-releasing agents, minimizes wastage of precious samples and reagents while handling, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Format Rack
Tip Style Automation
For Use With (Equipment) Beckman Coulter Multimek
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.