Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Universal Fit Micropipette Tips

Catalog No. 14222720 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Color:
Clear
Tip Style:
Low Retention, Beveled
Quantity:
4800/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Color Tip Style Quantity
14-222-720 Non-sterile Clear Low Retention, Beveled 4800/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-720 Supplier Axygen™ Supplier No. T200CSTKS
Add to Cart
Add to Cart

Axygen™ Universal Fit Pipette Tips are compatible with most research-grade single- and multi-channel micropipettes, and are certified to be DNase-/RNase-free and nonpyrogenic.

  • RNase-/DNase free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Available in a variety of sizes
  • Available with a variety of tip styles including beveled, fine point, and wide bore
  • Available sterile/non-sterile
  • Available in bulk, racked, or stack rack packaging
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery

Order Info

CapacityMetric

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims Dnase-, Rnase-, Pyrogen-, Human gDNA-, PCR-Inhibition-Free
Features Maxymum Recovery™
Format Stack Rack
Tip Style Low Retention, Beveled
Packaging 96 x 5 Racks/Stack, 10 Stacks/Cs.
Quantity 4800/Cs.
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.