Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Slicone 96 Round Well Compression Flat Mat for PCR Microplates

Catalog No. 14222064 Shop All Axygen Products
Change view
Click to view available options
Material:
Silicone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
14-222-064 Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-064 Supplier Axygen™ Supplier No. CMFLAT
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suited to help reduce evaporation and ensure proper seal when used in conjunction with Axygen PCR Microplates and PlateMax™ sealing films

Axygen™ Compression Mats are ideally suited to help reduce evaporation and ensure a proper seal when used in conjunction with Axygen PCR Microlates and PlateMaxTM sealing films. Made of inert 100% virgin silicone, they are capable of withstanding numerous PCR cycles.

  • For use with 96/384 well and PCR microplates
  • TRUSTED_SUSTAINABILITY

    Specifications

    Color Natural
    Material Silicone
    Sterility Non-sterile
    Autoclavable Autoclavable
    No. of Wells 96
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Product Type Compression Mat
    Well Shape Round
    Plate Compatibility 96/384-well PCR Microplate
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.