Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Tecan™ Evo MCA96 Compatible Nested Pipet Tips
GREENER_CHOICE

Catalog No. 14223552 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
50 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Gray
Orange
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Color
14-223-552 200 μL Non-sterile Gray
14-223-553 200 μL Sterile Gray
14-223-551 50 μL Sterile Orange
14-223-550 50 μL Non-sterile Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-223-552 Supplier Axygen™ Supplier No. EV200NTR
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

768 tips for use with Tecan Evo MCA96 multi-channel arm

  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: MCA96
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Sterility Non-sterile
Color Gray
Tip Style Round
Packaging Nested tip rack
Quantity 40 Racks/Cs.
For Use With (Equipment) Tecan EVO MCA96
Product Line Tecan
Material Polypropylene
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.