Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Catalog No. 14222021 Shop All Axygen Products
Change view
Click to view available options
Pre-slit:
No
Yes
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pre-slit Sterility
14-222-021 No Non-sterile
14-221-903 Yes Non-sterile
14-222-022 No Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-222-021 Supplier Axygen™ Supplier No. AM500ULRD
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50/Cs.
No. per Pack 10/Pk.
Pre-slit No
Product Type Sealing Mat
Well Shape Round
Plate Compatibility 96-well Deep Well Plate, 0.5 mL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov