Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Axypet™ 12-Channel Pipettor

Axygen™ Axypet™ pipettors combine high levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.

$582.50

Specifications

Type Multi-Channel Pipettor
Warranty 3 years
Number of Channels 12
View More Specs

Products 4
Catalog Number Mfr. No. Volume (Metric) Accuracy Precision Description Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Accuracy Precision Description Price Quantity & Availability  
14-223-564
View Documents
Axygen™
AP1210
1 to 10 µL ±8.0/ ±2.0% ±6.0/ ±1.2% Tip Type: 10 µL
Case for $582.50
Add to cart
 
14-223-567
View Documents
Axygen™
AP1250
5 to 50 µL ±4.0/ ±1.6% ±2.5/ ±0.6% Tip Type: 200 µL
Case for $582.50
Add to cart
 
14-223-565
View Documents
Axygen™
AP12200
20 to 200 µL ±3.0/ ±1.0% ±1.5/ ±0.6% Tip Type: 200 µL
Case for $582.50
Add to cart
 
14-223-566
View Documents
Axygen™
AP12300
50 to 300 µL ±1.6/ ±1.0% ±1.5/ ±0.6% Tip Type: 300 µL
Case for $582.50
Add to cart
 
Description

Description

  • Thumb-friendly soft spring pipetting system reduces fatigue
  • 3-year warranty
  • Unique side EZIject button option reduces thumb movement
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
  • Each channel of the pipettor has an individual, precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next
  • Tip manifold rotates 360°
  • Shafts withdraw when pressed against a row of tips
  • Each shaft moves independently, minimizing jamming of tips
  • Micrometer is continuously adjustable for selection of whole or fractional volumes

Specifications

Specifications

Multi-Channel Pipettor
3 years
12
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit