Learn More
Corning Costar™ 96-well Tissue Culture Treated Plate
GREENER_CHOICE
Catalog No. 01000821 Shop All Corning Products
Change view
Click to view available options
96-well Tissue Culture Treated plate
Catalog No.
01-000-821 96-well Tissue Culture Treated plate
1 options
This item is not returnable. View return policy
Catalog No. 01-000-821 Supplier Corning Supplier No. 9167BC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Round bottom, with low evaporation lid, with barcode label, non-pyrogenic.
  • Virgin Polystyrene meets UPS, Class VI requirements for plastic containers and closures
  • Barcode label (format 128) on H12 short side centered
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tissue Culture Treated plate
WARNING: Cancer - www.P65Warnings.ca.gov