Learn More
Description
Utilizes magnetic bead technology for high-throughput purification of plasmid DNA from E. coli cells. AxyPrep MAG Plasmid Kit can also be used with fosmid and BAC vector-based constructs ranging from 5 Kb to 150 Kb.
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| Sample Type | E. coli Culture |
| For Use With (Application) | High throughput purification of plasmid DNA |
| No. of Reactions | 10 Preps |
| Product Type | Plasmid Purification Kit |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.