Learn More
Description
For isolation of genomic DNA from Rodent tail tissues. This protocol provides instruction to extract gDNA from fresh or frozen rodent tails.
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| Sample Type | Rodent Tail Tissue (Frozen or Fresh) |
| For Use With (Application) | To isolate genomic DNA |
| No. of Reactions | 10 Preps |
| Product Type | DNA Purification Kit |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.