Learn More
Description
The AxyPrep MAG Tissue-Blood gDNA Kit utilizes a magnetic beads-based technology to isolate genomic DNA from rodent tail tissues. This protocol provides instruction to extract gDNA from fresh or frozen rodent tails. The protocol is performed in 96-well format and 1.5 mL tubes. Purification begins by the addition of lysis buffer, and Proteinase K to rupture cell membranes and digest protein. Genomic DNA is then immobilized on magnetic particles by the addition of a magnetic binding reagent. This differential binding allows the gDNA to be easily separated from contaminants using a magnetic field. Contaminants can then be rinsed away using a simple washing procedure.
- Expanded protocol for animal tissue, cultured cells, buccal swabs, and blood
Specifications
Specifications
| Format | 96-well Plate, Tube |
| Isolation Technology | Magnetic Bead |
| For Use With (Application) | Isolation of genomic DNA from Rodent tail tissues |
| Includes | 125 mL TB1 Buffer, 120 mL TB2 Buffer, 88 mL Wash Buffer, 10 mL Proteinase K solution, 4.4 mL FB particles, 120 mL Elution Buffer, 1 protocol |
| No. of Reactions | 384 Preps |
| Product Type | Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.