Learn More
Description
- Cells are tumorigenic and are characterized by low adenylate cyclase activity. lack of FasL expression, and positive staining for the urothelial marker cytokeratin-7.5
- Tumors derived from AY-27 cells demonstrate orthotopic growth, allowing exposure to anti-tumor drugs in a natural environment
Specifications
Specifications
| Content And Storage | Store in liquid nitrogen, Cells can be cultured for 10 passages after initial thawing without significantly affecting cell marker expression and functionality |
| Quantity | 1 x 10&sup6; Cells/Vial |
| For Use With (Application) | Cell Culture |
| Sample Type | Cancer cells |
| Form | Liquid |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.