Learn More
Medchemexpress LLC AY 9944 100mg | 366-93-8 | 100MG
SDP
Catalog No. 5000239875
Encompass_Preferred
AY 9944 is a specific cholesterol biosynthesis inhibitor AY 9944 inhibits the 7-dehydro cholesterol 7-reductase (DHCR7) enzyme with an IC50 of 13 nM AY 9944 causes hypocholesterolemia and accumulation of 7DHC At high doses AY 9944 inhibits also in cultured embryos sterol 7- 8 isomerase which causes the accumulation of cholest-8-en-3 -ol[1 [2 [3
Catalog No. 50-002-39875 Supplier Medchemexpress LLC Supplier No. HY107420100MG
AY 9944 is a specific cholesterol biosynthesis inhibitor AY 9944 inhibits the 7-dehydro cholesterol 7-reductase (DHCR7) enzyme with an IC50 of 13 nM AY 9944 causes hypocholesterolemia and accumulation of 7DHC At high doses AY 9944 inhibits also in cultured embryos sterol 7- 8 isomerase which causes the accumulation of cholest-8-en-3 -ol[1 [2 [3