Learn More
Medchemexpress LLC Ay 9944 | 366-93-8 | 99.7% | 464.30 | C22H30Cl4N2 | 25 MG
SDP
Catalog No. 5000239779
Encompass_Preferred
AY 9944 is a potent small-molecule inhibitor of 7-dehydrocholesterol Δ7-reductase (DHCR7) used to block cholesterol biosynthesis in research. It inhibits DHCR7 with an IC50 of approximately 13 nM, causing accumulation of 7-dehydrocholesterol and affecting sterol metabolism and signaling pathways.
Catalog No. 50-002-39779 Supplier Medchemexpress LLC Supplier No. HY10742025MG
AY 9944 is a potent small-molecule inhibitor of 7-dehydrocholesterol Δ7-reductase (DHCR7) used to block cholesterol biosynthesis in research. It inhibits DHCR7 with an IC50 of approximately 13 nM, causing accumulation of 7-dehydrocholesterol and affecting sterol metabolism and signaling pathways.