Learn More
Medchemexpress LLC Ay 9944 | 366-93-8 | MFCD00034973 | 99.7% | 464.30 g/mol | C22H30Cl4N2 | 50 MG
SDP
Catalog No. 5000239883
Encompass_Preferred
AY 9944 is a small-molecule inhibitor of 7-dehydrocholesterol Δ7-reductase (DHCR7) used in research to block cholesterol biosynthesis and probe sterol metabolism and signaling pathways.
  • Potent DHCR7 inhibitor (IC50 ≈ 13 nM).
  • Induces accumulation of 7-dehydrocholesterol and reduces cholesterol synthesis.
  • Supplied as a dihydrochloride salt for stability.
  • High purity suitable for biochemical and cell-based studies.
  • Soluble in DMSO and compatible with aqueous buffers.
  • Used to study cholesterol-related pathways and hedgehog signaling.
Catalog No. 50-002-39883 Supplier Medchemexpress LLC Supplier No. HY10742050MG
Add to Cart
Add to Cart
AY 9944 is a small-molecule inhibitor of 7-dehydrocholesterol Δ7-reductase (DHCR7) used in research to block cholesterol biosynthesis and probe sterol metabolism and signaling pathways.
  • Potent DHCR7 inhibitor (IC50 ≈ 13 nM).
  • Induces accumulation of 7-dehydrocholesterol and reduces cholesterol synthesis.
  • Supplied as a dihydrochloride salt for stability.
  • High purity suitable for biochemical and cell-based studies.
  • Soluble in DMSO and compatible with aqueous buffers.
  • Used to study cholesterol-related pathways and hedgehog signaling.