Learn More
Medchemexpress LLC AZ 11645373 | 227088-94-0 | 99.54% | 463.51 | 1 ML
Supplier: Medchemexpress LLC HY1086701MG
AZ11645373 is a highly selective and potent antagonist at human but not rat P2X7 receptors. It inhibits ATP-evoked IL-1β release from lipopolysaccharide-activated THP-1 cells, with an IC50 value of 90 nM.
- Highly selective and potent antagonist at human P2X7 receptors.
- Inhibits ATP-evoked IL-1β release from lipopolysaccharide-activated THP-1 cells.
- Produces a concentration-dependent inhibition of BzATP-mediated calcium transients.
- No significant effect on basal levels of IL-1β in culture medium of LPS-treated cells at specified concentrations.