Learn More
Cayman Chemical AZ 13705339 50mg
SDP
Catalog No. 502945341
Encompass_Preferred
A PAK1 inhibitor (IC50 = 0.33 nM); selective for PAK1 over a panel of 117 kinases at 100 nM; binds to PAK1 and PAK2 (Kd = 0.28 and 0.32 nM, respectively); inhibits S. enterica infection in HAP1 cells at 10 µM; inhibits cell death induced by an antibody targeting Siglec-8 in IL-5-stimulated primary human eosinophils (IC50 = 59 nM)
Catalog No. 50-294-5341 Supplier Cayman Chemical Supplier No. 4208750 MG
A PAK1 inhibitor (IC50 = 0.33 nM); selective for PAK1 over a panel of 117 kinases at 100 nM; binds to PAK1 and PAK2 (Kd = 0.28 and 0.32 nM, respectively); inhibits S. enterica infection in HAP1 cells at 10 µM; inhibits cell death induced by an antibody targeting Siglec-8 in IL-5-stimulated primary human eosinophils (IC50 = 59 nM)