Learn More
Abcam AZ-20, 25MG
Catalog No. 502671860
Encompass_Preferred
MW 412.5 Da. Potent and selective ATR kinase inhibitor (IC₅₀ = 5 nM in vitro; IC₅₀ = 50 nM in carcinoma cell lines). Selective versus mTOR (8-fold). Displayed selectivity versus a panel of 442 kinases.Inhibits LoVo colorectal adenocarcinoma tumor cell growth.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1860 Supplier Abcam Supplier No. AB27362325MG
MW 412.5 Da. Potent and selective ATR kinase inhibitor (IC₅₀ = 5 nM in vitro; IC₅₀ = 50 nM in carcinoma cell lines). Selective versus mTOR (8-fold). Displayed selectivity versus a panel of 442 kinases.Inhibits LoVo colorectal adenocarcinoma tumor cell growth.
The product is subject to the following: Abcam Restricted Use Statement