Learn More
Medchemexpress LLC AZ-3137 | 2960179-55-7 | 99.9% | 801.87 | 5 MG
SDP
Catalog No. 5000393946
Encompass_Preferred
AZ'3137 is an orally active PROTAC-type androgen receptor (AR) degrader with a DC50 value of 22 nM. It can degrade L702H mutant AR (DC50 of 92 nM) and inhibit cell proliferation of LNCaP, with a GI50 value of 74 nM. It can also inhibit AR signaling and tumor growth in prostate cancer mice.
  • Orally active PROTAC-type androgen receptor (AR) degrader.
  • Degrades L702H mutant AR.
  • Inhibits cell proliferation of LNCaP cells.
  • Inhibits AR signaling and tumor growth in prostate cancer mice.
  • Inhibits AR signaling and cell proliferation of prostate cancer cell line LNCaP (0-10 μM; 7 days).
  • Inhibits the expression of AR target genes in LNCaP cells (0.1 nM-10 μM; 24 h) and degrades AR in C4-2 and VCAP cells (0.1 nM-10 μM; 24 h).
  • Inhibits tumor growth in prostate cancer mice (oral gavage (p.o.); once daily; for 10 days).
Catalog No. 50-003-93946 Supplier Medchemexpress LLC Supplier No. HY1627025MG
AZ'3137 is an orally active PROTAC-type androgen receptor (AR) degrader with a DC50 value of 22 nM. It can degrade L702H mutant AR (DC50 of 92 nM) and inhibit cell proliferation of LNCaP, with a GI50 value of 74 nM. It can also inhibit AR signaling and tumor growth in prostate cancer mice.
  • Orally active PROTAC-type androgen receptor (AR) degrader.
  • Degrades L702H mutant AR.
  • Inhibits cell proliferation of LNCaP cells.
  • Inhibits AR signaling and tumor growth in prostate cancer mice.
  • Inhibits AR signaling and cell proliferation of prostate cancer cell line LNCaP (0-10 μM; 7 days).
  • Inhibits the expression of AR target genes in LNCaP cells (0.1 nM-10 μM; 24 h) and degrades AR in C4-2 and VCAP cells (0.1 nM-10 μM; 24 h).
  • Inhibits tumor growth in prostate cancer mice (oral gavage (p.o.); once daily; for 10 days).