Learn More
Selleck Chemical LLC AZ 3146 10mg 1124329-14-1
SDP
Supplier:  Selleck Chemical LLC S273110MG
Encompass_Preferred
AZ3146 is a selective Mps1 inhibitor with IC50 of ~35 nM, contributes to recruitment of CENP-E (kinesin-related motor protein), less potent to FAK, JNK1, JNK2, and Kit. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2863