Learn More
Selleck Chemical LLC AZ 3146 S2731-10mM/1mL
SDP
Catalog No. 503112285
Encompass_Preferred
AZ3146 is a selective Mps1 inhibitor with IC50 of 35 nM contributes to recruitment of CENP-E (kinesin-related motor protein) less potent to FAK JNK1 JNK2 and Kit
Catalog No. 50-311-2285 Supplier Selleck Chemical LLC Supplier No. S273110MM/1ML
AZ3146 is a selective Mps1 inhibitor with IC50 of 35 nM contributes to recruitment of CENP-E (kinesin-related motor protein) less potent to FAK JNK1 JNK2 and Kit