Learn More
Selleck Chemical LLC AZ 628 S2746-1g
SDP
Catalog No. 503112469
Encompass_Preferred
AZ628 is a new pan-Raf inhibitor for BRAF BRAFV600E and c-Raf-1 with IC50 of 105 nM 34 nM and 29 nM in cell-free assays also inhibits VEGFR2 DDR2 Lyn Flt1 FMS etc AZ628 induces apoptosis
Catalog No. 50-311-2469 Supplier Selleck Chemical LLC Supplier No. S27461G
AZ628 is a new pan-Raf inhibitor for BRAF BRAFV600E and c-Raf-1 with IC50 of 105 nM 34 nM and 29 nM in cell-free assays also inhibits VEGFR2 DDR2 Lyn Flt1 FMS etc AZ628 induces apoptosis