Learn More
TARGETMOL CHEMICALS INC AZ 628 25MG
SDP
Catalog No. 502579435
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ628 is a new pan-Raf inhibitor for BRAF BRAFV600E and c-Raf-1 with IC50 of 105 nM 34 nM and 29 nM also inhibits VEGFR2 DDR2 Lyn Flt1 FMS etc. purity: 98%
Catalog No. 50-257-9435 Supplier TARGETMOL CHEMICALS INC Supplier No. T631825MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ628 is a new pan-Raf inhibitor for BRAF BRAFV600E and c-Raf-1 with IC50 of 105 nM 34 nM and 29 nM also inhibits VEGFR2 DDR2 Lyn Flt1 FMS etc. purity: 98%