Learn More
Medchemexpress LLC AZ 628 | 878739-06-1 | 99.4% | 451.52 | 50 MG
SDP
Catalog No. 5000344885
Encompass_Preferred
AZ 628 is a pan-Raf kinase inhibitor, effectively targeting B-Raf, B-RafV600E, and c-Raf-1. It also inhibits the activation of various tyrosine protein kinases, including VEGFR2, DDR2, Lyn, Flt1, and FMS. This compound is known to inhibit anchorage-dependent and -independent growth, induce cell cycle arrest, and promote apoptosis in colon and melanoma cell lines with the B-RafV600E mutation. Furthermore, it suppresses growth in cells expressing K-RASG13D and specifically affects the viability of K-RAS mutant cells.
  • Pan-Raf kinase inhibitor
  • Inhibits B-Raf, B-RafV600E, and c-Raf-1
  • Inhibits various tyrosine protein kinases
  • Inhibits anchorage-dependent and -independent growth
  • Causes cell cycle arrest
  • Induces apoptosis in colon and melanoma cell lines with B-RafV600E mutation
  • Suppresses growth in K-RASG13D expressing cells
  • Selectively affects viability in K-RAS mutant cells
Catalog No. 50-003-44885 Supplier Medchemexpress LLC Supplier No. HY1100410MM/1ML
Add to Cart
Add to Cart
AZ 628 is a pan-Raf kinase inhibitor, effectively targeting B-Raf, B-RafV600E, and c-Raf-1. It also inhibits the activation of various tyrosine protein kinases, including VEGFR2, DDR2, Lyn, Flt1, and FMS. This compound is known to inhibit anchorage-dependent and -independent growth, induce cell cycle arrest, and promote apoptosis in colon and melanoma cell lines with the B-RafV600E mutation. Furthermore, it suppresses growth in cells expressing K-RASG13D and specifically affects the viability of K-RAS mutant cells.
  • Pan-Raf kinase inhibitor
  • Inhibits B-Raf, B-RafV600E, and c-Raf-1
  • Inhibits various tyrosine protein kinases
  • Inhibits anchorage-dependent and -independent growth
  • Causes cell cycle arrest
  • Induces apoptosis in colon and melanoma cell lines with B-RafV600E mutation
  • Suppresses growth in K-RASG13D expressing cells
  • Selectively affects viability in K-RAS mutant cells