Learn More
Cayman Chemical AZ 7550 (hydrochloride), Purity: ≥98%, 5 mg
SDP
Catalog No. 502945329
Encompass_Preferred
An active metabolite of AZ 9291; inhibits IGF-1R in a cell-free assay (IC50 = 1.6 µM); inhibits EGFR autophosphorylation in H1975 cells expressing EGFRT790M/L858R, PC-9 cells expressing EGFRExon19del, and LoVo cells expressing wild-type EGFR (IC50s = 0.045, 0.026, and 0.786 µM, respectively)
Catalog No. 50-294-5329 Supplier Cayman Chemical Supplier No. 407535 MG
An active metabolite of AZ 9291; inhibits IGF-1R in a cell-free assay (IC50 = 1.6 µM); inhibits EGFR autophosphorylation in H1975 cells expressing EGFRT790M/L858R, PC-9 cells expressing EGFRExon19del, and LoVo cells expressing wild-type EGFR (IC50s = 0.045, 0.026, and 0.786 µM, respectively)