Learn More
Medchemexpress LLC AZ'9567 | 3056570-19-2 | 99.2% | 447.44 | 1 MG
SDP
Catalog No. 5000392983
Encompass_Preferred
AZ'9567 is a potent MAT2A inhibitor that exhibits antiproliferative activity against MTAP KO HCT116 cells with a pIC50 of 8.9. This compound is supplied as a solid, white to off-white in appearance, and is soluble in DMSO.
  • Potent MAT2A inhibitor.
  • Exhibits antiproliferative activity against MTAP KO HCT116 cells with a pIC50 of 8.9.
  • Supplied as a solid.
  • White to off-white in appearance.
  • Soluble in DMSO.
Catalog No. 50-003-92983 Supplier Medchemexpress LLC Supplier No. HY1623531MG
AZ'9567 is a potent MAT2A inhibitor that exhibits antiproliferative activity against MTAP KO HCT116 cells with a pIC50 of 8.9. This compound is supplied as a solid, white to off-white in appearance, and is soluble in DMSO.
  • Potent MAT2A inhibitor.
  • Exhibits antiproliferative activity against MTAP KO HCT116 cells with a pIC50 of 8.9.
  • Supplied as a solid.
  • White to off-white in appearance.
  • Soluble in DMSO.