Learn More
TARGETMOL CHEMICALS INC AZ PFKFB3 26 10MG
SDP
Catalog No. 502577384
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ PFKFB3 26 is an effective and selective PFKFB3 inhibitor with an IC50 of 23 nM. The IC50s for PFKFB1 and PFKFB2 are 2.06 and 0.384 uM respectively. purity: 98%
Catalog No. 50-257-7384 Supplier TARGETMOL CHEMICALS INC Supplier No. T1436510MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ PFKFB3 26 is an effective and selective PFKFB3 inhibitor with an IC50 of 23 nM. The IC50s for PFKFB1 and PFKFB2 are 2.06 and 0.384 uM respectively. purity: 98%