Learn More
Medchemexpress LLC HY-101971 10mg Medchemexpress, AZ PFKFB3 26 CAS:1704740-52-2 Purity:>98%
SDP
Catalog No. 501967939
Encompass_Preferred
Medchemexpress, HY-101971 10mg AZ PFKFB3 26 CAS:1704740-52-2 AZ PFKFB3 26 is a potent and selective inhibitor of the metabolic kinase PFKFB3 with an IC50 of 23 nM. AZ PFKFB3 26 inhibits PFKFB1 and PFKFB2 with IC50s of 2.06 and 0.384 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7939 Supplier Medchemexpress LLC Supplier No. HY10197110MG
Medchemexpress, HY-101971 10mg AZ PFKFB3 26 CAS:1704740-52-2 AZ PFKFB3 26 is a potent and selective inhibitor of the metabolic kinase PFKFB3 with an IC50 of 23 nM. AZ PFKFB3 26 inhibits PFKFB1 and PFKFB2 with IC50s of 2.06 and 0.384 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.