Learn More
Medchemexpress LLC Az PFKFB3 26 | 1704740-52-2 | MFCD30742931 | 98.3% | 402.49 | C24H26N4O2 | 25 MG
SDP
Catalog No. 5000248714
Encompass_Preferred
AZ PFKFB3 26 is a potent, selective small-molecule inhibitor of PFKFB3 used in research on glycolytic regulation and cancer metabolism. It displays low-nanomolar biochemical potency and measurable cellular effects in tumor cell assays, and is provided as a high-purity solid for laboratory use.
  • Potent PFKFB3 biochemical inhibitor (IC50 23 nM).
  • Selective over PFKFB1 and PFKFB2 (IC50s 2.06 μM and 0.384 μM).
  • Active in A549 cells (fructose-1,6-bisphosphate reduction IC50 0.343 μM).
  • High-purity research-grade material suitable for in vitro studies.
  • Available in small pack sizes for preclinical experiments.
Catalog No. 50-002-48714 Supplier Medchemexpress LLC Supplier No. HY10197125MG
AZ PFKFB3 26 is a potent, selective small-molecule inhibitor of PFKFB3 used in research on glycolytic regulation and cancer metabolism. It displays low-nanomolar biochemical potency and measurable cellular effects in tumor cell assays, and is provided as a high-purity solid for laboratory use.
  • Potent PFKFB3 biochemical inhibitor (IC50 23 nM).
  • Selective over PFKFB1 and PFKFB2 (IC50s 2.06 μM and 0.384 μM).
  • Active in A549 cells (fructose-1,6-bisphosphate reduction IC50 0.343 μM).
  • High-purity research-grade material suitable for in vitro studies.
  • Available in small pack sizes for preclinical experiments.