Learn More
Medchemexpress LLC AZ PFKFB3 26 | 1704740-52-2 | MFCD30742931 | 99.2% | 402.49 g·mol-1 | C24H26N4O2 | 50 MG
SDP
Catalog No. 5000246968
Encompass_Preferred
AZ PFKFB3 26 is a research-grade small-molecule inhibitor of PFKFB3 (CAS 1704740-52-2) supplied as a powder for in vitro and preclinical studies. The compound has formula C24H26N4O2 and a molecular weight of 402.49 g·mol-1. Analytical data and handling information are provided to support experimental use.
  • Documented purity available (COA shows ~99.2%).
  • Suitable for in vitro studies; soluble in DMSO at ~5 mg/mL.
  • Analytical data provided: COA, H NMR, LCMS, and SDS.
  • Storage conditions specified for powder and solutions to preserve stability.
  • Available in multiple research sizes, including 50 mg.
Catalog No. 50-002-46968 Supplier Medchemexpress LLC Supplier No. HY10197150MG
AZ PFKFB3 26 is a research-grade small-molecule inhibitor of PFKFB3 (CAS 1704740-52-2) supplied as a powder for in vitro and preclinical studies. The compound has formula C24H26N4O2 and a molecular weight of 402.49 g·mol-1. Analytical data and handling information are provided to support experimental use.
  • Documented purity available (COA shows ~99.2%).
  • Suitable for in vitro studies; soluble in DMSO at ~5 mg/mL.
  • Analytical data provided: COA, H NMR, LCMS, and SDS.
  • Storage conditions specified for powder and solutions to preserve stability.
  • Available in multiple research sizes, including 50 mg.