Learn More
Medchemexpress LLC Az-PFKFB3-67 | 1704741-11-6 | MFCD30536361 | 97.0% | 455.51 | C26H25N5O3 | 10 MG
SDP
Catalog No. 5000249164
Encompass_Preferred
A potent, selective small-molecule inhibitor of PFKFB3 for biochemical and cellular research. It is supplied as a white to off-white solid (C26H25N5O3, 455.51 g/mol) with reported purity of ~97% and high solubility in DMSO, intended for in vitro studies of glycolytic regulation.
  • Potent inhibition of PFKFB3 (IC50 11 nM).
  • Selective activity versus related isoforms (PFKFB2 IC50 159 nM; PFKFB1 IC50 1130 nM).
  • Suitable for biochemical and cellular assays.
  • High solubility in DMSO, enabling concentrated stock solutions.
  • Stable as a powder at -20°C for long-term storage.
  • Available in small research pack sizes for assay development.
Catalog No. 50-002-49164 Supplier Medchemexpress LLC Supplier No. HY10197210MG
A potent, selective small-molecule inhibitor of PFKFB3 for biochemical and cellular research. It is supplied as a white to off-white solid (C26H25N5O3, 455.51 g/mol) with reported purity of ~97% and high solubility in DMSO, intended for in vitro studies of glycolytic regulation.
  • Potent inhibition of PFKFB3 (IC50 11 nM).
  • Selective activity versus related isoforms (PFKFB2 IC50 159 nM; PFKFB1 IC50 1130 nM).
  • Suitable for biochemical and cellular assays.
  • High solubility in DMSO, enabling concentrated stock solutions.
  • Stable as a powder at -20°C for long-term storage.
  • Available in small research pack sizes for assay development.