Learn More
TARGETMOL CHEMICALS INC AZ084 5MG
SDP
Catalog No. 502581723
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ084 is a potent selective allosteric and oral active CCR8 antagonist (Ki 0.9 nM). It has the potential to treat asthma. purity: 98%
Catalog No. 50-258-1723 Supplier TARGETMOL CHEMICALS INC Supplier No. T104255MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AZ084 is a potent selective allosteric and oral active CCR8 antagonist (Ki 0.9 nM). It has the potential to treat asthma. purity: 98%